CH4005994546 ChemBridge Corporation

5-amino-2-(tetrahydro-2-furanylmethyl)-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD02767207
  • Cas Number : 434297-53-7
  • Molecular Weight : 246.2619

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7045775

Safety Information

MSDS https://www.hit2lead.com/sds/7045775_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart