CH4005996136 ChemBridge Corporation

N-(3-bromo-4-chlorophenyl)-2-(4-ethylphenoxy)acetamide

  • MDL number MFCD03554579
  • Cas Number : 527746-29-8
  • Molecular Weight : 368.6528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7086099

Safety Information

MSDS https://www.hit2lead.com/sds/7086099_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart