CH4005996162 ChemBridge Corporation

N-(4-bromo-2-isopropylphenyl)-2-(4-ethylphenoxy)acetamide

  • MDL number MFCD03555609
  • Cas Number : 528531-90-0
  • Molecular Weight : 376.2875

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7087119

Safety Information

MSDS https://www.hit2lead.com/sds/7087119_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart