CH4005997779 ChemBridge Corporation

[(4-ethyl-7-methyl-2-oxo-2H-chromen-5-yl)oxy]acetic acid

  • MDL number MFCD03713023
  • Cas Number : 690681-33-5
  • Molecular Weight : 262.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7105116

Safety Information

MSDS https://www.hit2lead.com/sds/7105116_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart