CH4005998056 ChemBridge Corporation

5-hydroxy-3,4,7-trimethyl-2H-chromen-2-one

  • MDL number MFCD03713196
  • Cas Number : 107057-96-5
  • Molecular Weight : 204.2219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7109536

Safety Information

MSDS https://www.hit2lead.com/sds/7109536_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart