CH4005998882 ChemBridge Corporation

2-(4-bromophenyl)-1-(2,4-dihydroxyphenyl)ethanone

  • MDL number MFCD00499092
  • Cas Number : 92152-60-8
  • Molecular Weight : 307.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7113606

Safety Information

MSDS https://www.hit2lead.com/sds/7113606_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart