CH4005999058 ChemBridge Corporation

3-amino-3-(4-ethoxy-1-naphthyl)propanoic acid

  • MDL number MFCD03001189
  • Cas Number : 612047-63-9
  • Molecular Weight : 259.3004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7114224

Safety Information

MSDS https://www.hit2lead.com/sds/7114224_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart