CH4006010024 ChemBridge Corporation

3-bromo-N-cyclopropylbenzamide

  • MDL number MFCD03387330
  • Cas Number : 337535-74-7
  • Molecular Weight : 240.0965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7223774

Safety Information

MSDS https://www.hit2lead.com/sds/7223774_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart