CH4006021781 ChemBridge Corporation

2-(butyrylamino)benzoic acid

  • MDL number MFCD00448228
  • Cas Number : 6328-94-5
  • Molecular Weight : 207.2258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7356177

Safety Information

MSDS https://www.hit2lead.com/sds/7356177_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart