CH4006022040 ChemBridge Corporation

1-(3-chlorobenzoyl)piperidine-4-carboxylic acid

  • MDL number MFCD03015383
  • Cas Number : 401581-33-7
  • Molecular Weight : 267.7082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7357137

Safety Information

MSDS https://www.hit2lead.com/sds/7357137_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart