CH4006022686 ChemBridge Corporation

2-[(2-phenylethyl)thio]nicotinic acid

  • MDL number MFCD03621206
  • Cas Number : 496775-95-2
  • Molecular Weight : 259.3235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7360080

Safety Information

MSDS https://www.hit2lead.com/sds/7360080_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart