CH4006026731 ChemBridge Corporation

2-(allyloxy)-5-bromobenzaldehyde

  • MDL number MFCD02257424
  • Cas Number : 40359-62-4
  • Molecular Weight : 241.0813

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7426395

Safety Information

MSDS https://www.hit2lead.com/sds/7426395_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart