CH4006027148 ChemBridge Corporation

N-(2-aminoethyl)-4-methoxybenzenesulfonamide

  • MDL number MFCD03634547
  • Cas Number : 90566-22-6
  • Molecular Weight : 230.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7436664

Safety Information

MSDS https://www.hit2lead.com/sds/7436664_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart