CH4014053366 ChemBridge Corporation

4-[(2,5-dichlorophenyl)sulfonyl]butan-2-one

  • MDL number MFCD00450449
  • Cas Number : 336195-37-0
  • Molecular Weight : 281.1556

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5230096

Safety Information

MSDS https://www.hit2lead.com/sds/5230096_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart