CH4014053971 ChemBridge Corporation

2-[methyl(2-thienylcarbonyl)amino]benzoic acid

  • MDL number MFCD00583358
  • Cas Number : 351000-22-1
  • Molecular Weight : 261.2963

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5257456

Safety Information

MSDS https://www.hit2lead.com/sds/5257456_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart