CH4014060633 ChemBridge Corporation

1-(2-hydroxy-4-methylphenyl)-3-(3-methylphenyl)propane-1,3-dione

  • MDL number MFCD20502991
  • Cas Number : 420823-80-9
  • Molecular Weight : 268.3071

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5811738

Safety Information

MSDS https://www.hit2lead.com/sds/5811738_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart