CH4014063054 ChemBridge Corporation

4-[(Z)-(4-oxo-2-thioxo-1,3-thiazolidin-5-ylidene)methyl]benzoic acid

  • MDL number MFCD01909747
  • Cas Number : 1287651-45-9
  • Molecular Weight : 265.3082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5913763

Safety Information

MSDS https://www.hit2lead.com/sds/5913763_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart