CH4014063494 ChemBridge Corporation

[3-(4-chlorophenyl)-1-phenyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD01909761
  • Cas Number : 36640-39-8
  • Molecular Weight : 284.7402

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5916616

Safety Information

MSDS https://www.hit2lead.com/sds/5916616_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart