CH4014063697 ChemBridge Corporation

(2E)-3-[3-(3-bromophenyl)-1-phenyl-1H-pyrazol-4-yl]acrylic acid

  • MDL number MFCD02633809
  • Cas Number : 372107-21-6
  • Molecular Weight : 369.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5918099

Safety Information

MSDS https://www.hit2lead.com/sds/5918099_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart