CH4014064609 ChemBridge Corporation

1-(4-bromophenyl)-2-[(4-chlorophenyl)amino]ethanone

  • MDL number MFCD01220318
  • Cas Number : 28012-58-0
  • Molecular Weight : 324.6002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5924139

Safety Information

MSDS https://www.hit2lead.com/sds/5924139_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart