CH4014064673 ChemBridge Corporation

1-(4-bromophenyl)-3-[(4-methylphenyl)amino]propan-1-one

  • MDL number MFCD01220247
  • Cas Number : 37155-15-0
  • Molecular Weight : 318.2083

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5924424

Safety Information

MSDS https://www.hit2lead.com/sds/5924424_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart