CH4014065152 ChemBridge Corporation

1-(4-bromophenyl)-3-[(4-ethylphenyl)amino]propan-1-one

  • MDL number MFCD00423361
  • Cas Number : 423735-02-8
  • Molecular Weight : 332.2349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5927359

Safety Information

MSDS https://www.hit2lead.com/sds/5927359_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart