CH4014067827 ChemBridge Corporation

4-chloro-3-(2,5-dimethyl-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD01762487
  • Cas Number : 26165-62-8
  • Molecular Weight : 249.6929

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5949510

Safety Information

MSDS https://www.hit2lead.com/sds/5949510_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart