CH4014068174 ChemBridge Corporation

3-(2,5-dimethyl-1H-pyrrol-1-yl)-2-methylbenzoic acid

  • MDL number MFCD02085007
  • Cas Number : 83141-00-8
  • Molecular Weight : 229.2744

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5953822

Safety Information

MSDS https://www.hit2lead.com/sds/5953822_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart