CH4014068460 ChemBridge Corporation

(2E)-3-(3-bromo-4-ethoxy-5-methoxyphenyl)acrylic acid

  • MDL number MFCD01873769
  • Cas Number : 354531-46-7
  • Molecular Weight : 301.1332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5957528

Safety Information

MSDS https://www.hit2lead.com/sds/5957528_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart