CH4014068611 ChemBridge Corporation

(2E)-3-(3-ethoxy-5-iodo-4-methoxyphenyl)acrylic acid

  • MDL number MFCD01873777
  • Cas Number : 340216-06-0
  • Molecular Weight : 348.1337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5959502

Safety Information

MSDS https://www.hit2lead.com/sds/5959502_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart