CH4014068756 ChemBridge Corporation

6-(4-bromophenyl)-2,3-dihydroimidazo[2,1-b][1,3]thiazole

  • MDL number MFCD00268935
  • Cas Number : 35181-52-3
  • Molecular Weight : 281.1716

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5961326

Safety Information

MSDS https://www.hit2lead.com/sds/5961326_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart