CH4014068911 ChemBridge Corporation

2-(4-mercaptophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD01604600
  • Cas Number : 332129-06-3
  • Molecular Weight : 299.3013

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5963135

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/5963135_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart