CH4014069270 ChemBridge Corporation

(2E)-3-(5-bromo-2,4-diethoxyphenyl)acrylic acid

  • MDL number MFCD02212564
  • Cas Number : 423752-65-2
  • Molecular Weight : 315.1598

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5967408

Safety Information

MSDS https://www.hit2lead.com/sds/5967408_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart