CH4014069289 ChemBridge Corporation

1-(2-hydroxy-5-methoxyphenyl)-3-phenylpropane-1,3-dione

  • MDL number MFCD02043576
  • Cas Number : 67029-87-2
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5967686

Safety Information

MSDS https://www.hit2lead.com/sds/5967686_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart