CH4014069373 ChemBridge Corporation

(2E)-3-(2-bromo-4-ethoxy-5-methoxyphenyl)acrylic acid

  • MDL number MFCD02212599
  • Cas Number : 423753-00-8
  • Molecular Weight : 301.1332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5968578

Safety Information

MSDS https://www.hit2lead.com/sds/5968578_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart