CH4014069641 ChemBridge Corporation

2-[5-(3-bromophenyl)-1H-pyrazol-3-yl]-5-methylphenol

  • MDL number MFCD20502999
  • Cas Number : 423753-91-7
  • Molecular Weight : 329.1912

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5971672

Safety Information

MSDS https://www.hit2lead.com/sds/5971672_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart