CH4014073365 ChemBridge Corporation

5-(4-bromo-2-fluorophenyl)-2-furaldehyde

  • MDL number MFCD03199045
  • Cas Number : 444284-83-7
  • Molecular Weight : 269.0665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6248865

Safety Information

MSDS https://www.hit2lead.com/sds/6248865_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart