CH4014075981 ChemBridge Corporation

2-bromo-3-chloro-5-ethoxy-4-hydroxybenzaldehyde

  • MDL number MFCD03923602
  • Cas Number : 597545-04-5
  • Molecular Weight : 279.515

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6484056

Safety Information

MSDS https://www.hit2lead.com/sds/6484056_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart