CH4014076883 ChemBridge Corporation

4-[5-(4-fluorophenyl)-2H-tetrazol-2-yl]butanoic acid

  • MDL number MFCD02932968
  • Cas Number : 384859-48-7
  • Molecular Weight : 250.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6541111

Safety Information

MSDS https://www.hit2lead.com/sds/6541111_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart