CH4014078653 ChemBridge Corporation

5,6-dimethyl-2-(2-propyn-1-ylthio)thieno[2,3-d]pyrimidin-4-amine

  • MDL number MFCD02329922
  • Cas Number : 315695-36-4
  • Molecular Weight : 249.3551

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6625466

Safety Information

MSDS https://www.hit2lead.com/sds/6625466_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart