CH4014079118 ChemBridge Corporation

3-amino-4-[(pyridin-3-ylmethyl)amino]benzoic acid

  • MDL number MFCD03274476
  • Cas Number : 436088-89-0
  • Molecular Weight : 243.2612

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6637326

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6637326
Availability details for
Quantity
  • Availability
  • Add to Cart