CH4014082436 ChemBridge Corporation

(5-oxo-2-phenylcyclopent-1-en-1-yl)acetic acid

  • MDL number MFCD01839088
  • Cas Number : 42882-19-9
  • Molecular Weight : 216.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6832223

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6832223
Availability details for
Quantity
  • Availability
  • Add to Cart