CH4014082634 ChemBridge Corporation

[(1-benzyl-1H-indol-3-yl)thio]acetic acid

  • MDL number MFCD03410208
  • Cas Number : 524035-97-0
  • Molecular Weight : 297.3715

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6848227

Safety Information

MSDS https://www.hit2lead.com/sds/6848227_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart