CH4014093805 ChemBridge Corporation

2-(4-formyl-2-methoxyphenoxy)ethyl acetate

  • MDL number MFCD02380339
  • Cas Number : 692275-82-4
  • Molecular Weight : 238.2366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7423058

Safety Information

MSDS https://www.hit2lead.com/sds/7423058_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart