CH4014094306 ChemBridge Corporation

4-(allyloxy)-2-chloro-5-methoxybenzaldehyde

  • MDL number MFCD02380262
  • Cas Number : 692279-00-8
  • Molecular Weight : 226.6562

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7433377

Safety Information

MSDS https://www.hit2lead.com/sds/7433377_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart