CH4017863714 ChemBridge Corporation

2-ethyl-3,6-dimethyltetrahydro-4H-thiopyran-4-one

  • MDL number MFCD00461807
  • Cas Number : 462065-51-6
  • Molecular Weight : 172.2877

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5106463

Safety Information

MSDS https://www.hit2lead.com/sds/5106463_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart