CH4017867321 ChemBridge Corporation

2-(2-nitrophenoxy)benzaldehyde

  • MDL number MFCD00156891
  • Cas Number : 66961-19-1
  • Molecular Weight : 243.2149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5144034

Safety Information

MSDS https://www.hit2lead.com/sds/5144034_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart