CH4017868492 ChemBridge Corporation

2-(2-hydroxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD00411806
  • Cas Number : 312746-96-6
  • Molecular Weight : 283.2357

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5160622

Safety Information

MSDS https://www.hit2lead.com/sds/5160622_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart