CH4017871500 ChemBridge Corporation

N-(5-chloro-2-pyridinyl)-3-oxobutanamide

  • MDL number MFCD00457344
  • Cas Number : 64500-19-2
  • Molecular Weight : 212.633

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5222291

Safety Information

MSDS https://www.hit2lead.com/sds/5222291_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart