CH4017874505 ChemBridge Corporation

N-1,3-benzodioxol-5-yl-3-oxobutanamide

  • MDL number MFCD00744554
  • Cas Number : 35493-00-6
  • Molecular Weight : 221.2093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5267750

Safety Information

MSDS https://www.hit2lead.com/sds/5267750_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart