CH4017875192 ChemBridge Corporation

N~3~-(2-phenylethyl)-beta-alaninamide

  • MDL number MFCD00769479
  • Cas Number : 4091-84-3
  • Molecular Weight : 192.2575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5301488

Safety Information

MSDS https://www.hit2lead.com/sds/5301488_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart