CH4017876775 ChemBridge Corporation

3-{[(methylthio)carbonyl]amino}adamantane-1-carboxylic acid

  • MDL number MFCD01830815
  • Cas Number : 342046-29-1
  • Molecular Weight : 269.3599

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5347086

Safety Information

MSDS https://www.hit2lead.com/sds/5347086_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart