CH4017878472 ChemBridge Corporation

2-[(4-methylphenoxy)methyl]-3-furoic acid

  • MDL number MFCD00442870
  • Cas Number : 331670-04-3
  • Molecular Weight : 232.232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5404455

Safety Information

MSDS https://www.hit2lead.com/sds/5404455_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart