CH4017892570 ChemBridge Corporation

methyl N-[1-(4-methoxyphenyl)-2,5-dioxopyrrolidin-3-yl]glycinate

  • MDL number MFCD01907601
  • Cas Number : 471917-79-0
  • Molecular Weight : 292.2872

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5855661

Safety Information

MSDS https://www.hit2lead.com/sds/5855661_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart